4-Iodo-1-tritylimidazole
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Iodo-1-trityl-1H-imidazole
019056-1G
019056-5G
FLUO019056-1GEA
0
GBP
FLUO019056-1G
FLUO019056-5G
4-Iodo-1-tritylimidazole
4-Iodo-1-tritylimidazole
Formula:
C₂₂H₁₇IN₂ |
MDL Number:
MFCD02179542 CAS Number: 96797-15-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...