3-(Trifluoromethyl)picolinic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20927G1 AS20927G5
ACTIAS20927G1EA 0 GBP
ACTIAS20927G1 ACTIAS20927G5
3-(Trifluoromethyl)picolinic acid
3-(Trifluoromethyl)picolinic acid
Formula: C₇H₄F₃NO₂
MDL Number: MFCD07375386
CAS Number: 87407-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR