1-(3-Chlorophenyl)piperidine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-6448-5G
HC-6448-1G
COBBHC-6448-5GEA
0
GBP
COBBHC-6448-5G
COBBHC-6448-1G
1-(3-Chlorophenyl)piperidine
1-(3-Chlorophenyl)piperidine
Formula:
C₁₁H₁₄ClN |
MDL Number:
MFCD10699658 CAS Number: 102563-84-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...