1-(3-Chloro-4-methylphenyl)piperidine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6384-5G HC-6384-1G
COBBHC-6384-5GEA 0 GBP
COBBHC-6384-5G COBBHC-6384-1G
1-(3-Chloro-4-methylphenyl)piperidine
1-(3-Chloro-4-methylphenyl)piperidine
Formula: C₁₂H₁₆ClN
MDL Number: MFCD09878387
CAS Number: 1000339-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR