1-(3-Chloro-2-methylphenyl)piperidine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-6489-5G
HC-6489-1G
COBBHC-6489-5GEA
0
GBP
COBBHC-6489-5G
COBBHC-6489-1G
1-(3-Chloro-2-methylphenyl)piperidine
1-(3-Chloro-2-methylphenyl)piperidine
Formula:
C₁₂H₁₆ClN |
MDL Number:
MFCD09972164 CAS Number: 1020253-08-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...