1-(2-Bromophenyl)piperidin-2-one
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OT-1239-1G
OT-1239-5G
COBBOT-1239-1GEA
0
GBP
COBBOT-1239-1G
COBBOT-1239-5G
1-(2-Bromophenyl)piperidin-2-one
1-(2-Bromophenyl)piperidin-2-one
Formula:
C₁₁H₁₂BrNO |
MDL Number:
MFCD04037062 CAS Number: 917508-51-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...