1-(3-Bromo-4-methylphenyl)piperidine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-2493-5G
HC-2493-1G
COBBHC-2493-5GEA
0
GBP
COBBHC-2493-5G
COBBHC-2493-1G
1-(3-Bromo-4-methylphenyl)piperidine
1-(3-Bromo-4-methylphenyl)piperidine
Formula:
C₁₂H₁₆BrN |
MDL Number:
MFCD11855915 CAS Number: 761001-65-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...