3-Nitropicolinic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS24072G1 AS24072G5
ACTIAS24072G1EA 0 GBP
ACTIAS24072G1 ACTIAS24072G5
3-Nitropicolinic acid
3-Nitropicolinic acid
Formula: C₆H₄N₂O₄
MDL Number: MFCD06658500
CAS Number: 59290-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR