2'-Nitro-3',4',6'-trifluoroacetanilide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-1384-5G AN-1384-1G
COBBAN-1384-5GEA 0 GBP
COBBAN-1384-5G COBBAN-1384-1G
2'-Nitro-3',4',6'-trifluoroacetanilide
2'-Nitro-3',4',6'-trifluoroacetanilide
Formula: C₈H₅F₃N₂O₃
MDL Number: MFCD11504834
CAS Number: 388-11-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR