8-(Trifluoromethyl)-4-quinolinol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Hydroxy-8-(trifluoromethyl)quinoline
007675-1G
007675-5G
FLUO007675-1GEA
0
GBP
FLUO007675-1G
FLUO007675-5G
8-(Trifluoromethyl)-4-quinolinol
8-(Trifluoromethyl)-4-quinolinol
Formula:
C₁₀H₆F₃NO |
MDL Number:
MFCD00134577 CAS Number: 23779-96-6 EINECS: 245-879-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...