8-(Trifluoromethyl)-4-quinolinol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Hydroxy-8-(trifluoromethyl)quinoline

AS20895G25 AS20895G5
ACTIAS20895G25EA 357 GBP
ACTIAS20895G25 ACTIAS20895G5
8-(Trifluoromethyl)-4-quinolinol
8-(Trifluoromethyl)-4-quinolinol
Formula: C₁₀H₆F₃NO
MDL Number: MFCD00134577
CAS Number: 23779-96-6
EINECS: 245-879-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR