4-Butylbiphenyl

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Butyl-1,1'-Biphenyl

AS25317G100 AS25317G25
ACTIAS25317G100EA 75 GBP
ACTIAS25317G100 ACTIAS25317G25
4-Butylbiphenyl
4-Butylbiphenyl
Formula: C₁₆H₁₈
MW: 210.32 g/mol
MDL Number: MFCD00100665
CAS Number: 37909-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR