5-Bromo-4-fluoro-2-methylaniline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS24821G100 AS24821G25 AS24821G5
ACTIAS24821G100EA 216 GBP
ACTIAS24821G100 ACTIAS24821G25 ACTIAS24821G5
5-Bromo-4-fluoro-2-methylaniline
5-Bromo-4-fluoro-2-methylaniline
Formula: C₇H₇BrFN
MW: 204.04 g/mol
MDL Number: MFCD05865218
CAS Number: 627871-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR