1-Bromo-4-(difluoromethyl)benzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Bromobenzal fluoride
023880-1G
023880-5G
FLUO023880-1GEA
0
GBP
FLUO023880-1G
FLUO023880-5G
1-Bromo-4-(difluoromethyl)benzene
1-Bromo-4-(difluoromethyl)benzene
Formula:
C₇H₅BrF₂ MW: 207.02 g/mol |
MDL Number:
MFCD06657964 CAS Number: 51776-71-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...