1-Bromo-3-(difluoromethyl)benzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Bromo-3-(difluoromethyl)benzene, 3-Bromobenzal fluoride

024453-1G 024453-5G
FLUO024453-1GEA 0 GBP
FLUO024453-1G FLUO024453-5G
1-Bromo-3-(difluoromethyl)benzene
1-Bromo-3-(difluoromethyl)benzene
Formula: C₇H₅BrF₂
MW: 207.015 g/mol
MDL Number: MFCD07368984
CAS Number: 29848-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR