4-Bromo-3-chlorophenylboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(4-Bromo-3-chlorophenyl)boronic acid
BB-6030-1G
BB-6030-5G
BB-6030-250M
COBBBB-6030-1GEA
0
GBP
COBBBB-6030-1G
COBBBB-6030-5G
COBBBB-6030-250M
4-Bromo-3-chlorophenylboronic acid
4-Bromo-3-chlorophenylboronic acid
Formula:
C₆H₅BBrClO₂ MW: 235.2700 g/mol |
MDL Number:
MFCD12913945 CAS Number: 1217501-28-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...