4-Bromo-2-chlorophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (4-Bromo-2-chlorophenyl)boronic acid

BB-5963-1G BB-5963-5G BB-5963-250M
COBBBB-5963-1GEA 0 GBP
COBBBB-5963-1G COBBBB-5963-5G COBBBB-5963-250M
4-Bromo-2-chlorophenylboronic acid
4-Bromo-2-chlorophenylboronic acid
Formula: C₆H₅BBrClO₂
MW: 235.2700 g/mol
MDL Number: MFCD13195646
CAS Number: 1046861-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR