2-Bromo-4-(trifluoromethyl)benzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Bromo-4-(trifluoromethyl)benzonitile

AS32355G1 AS32355G5
ACTIAS32355G1EA 0 GBP
ACTIAS32355G1 ACTIAS32355G5
2-Bromo-4-(trifluoromethyl)benzonitrile
2-Bromo-4-(trifluoromethyl)benzonitrile
Formula: C₈H₃BrF₃N
MW: 250.0200 g/mol
MDL Number: MFCD06659467
CAS Number: 35764-15-9
EINECS: 252-715-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR