4-Chloro-3-ethoxyaniline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AN-1353-5G
AN-1353-1G
COBBAN-1353-5GEA
0
GBP
COBBAN-1353-5G
COBBAN-1353-1G
4-Chloro-3-ethoxyaniline
4-Chloro-3-ethoxyaniline
Formula:
C₇H₈ClNO |
MDL Number:
MFCD11053831 CAS Number: 852854-42-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...