1-Bromo-4-(methoxymethoxy)benzene
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OT-1209-1G
OT-1209-5G
COBBOT-1209-1GEA
0
GBP
COBBOT-1209-1G
COBBOT-1209-5G
1-Bromo-4-(methoxymethoxy)benzene
1-Bromo-4-(methoxymethoxy)benzene
Formula:
C₈H₉BrO₂ MW: 217.0599 g/mol |
MDL Number:
MFCD00457268 CAS Number: 25458-45-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...