4-Fluoro-2-isopropoxyaniline hydrochloride
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AN-1038-5G
AN-1038-1G
COBBAN-1038-5GEA
0
GBP
COBBAN-1038-5G
COBBAN-1038-1G
4-Fluoro-2-isopropoxyaniline hydrochloride
4-Fluoro-2-isopropoxyaniline hydrochloride
Formula:
C₉H₁₃ClFNO MW: 205.6600 g/mol |
MDL Number:
MFCD01632194 CAS Number: 380430-47-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...