5-Fluoro-o-anisidine hydrochloride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-1332-1G AN-1332-25G AN-1332-5G
COBBAN-1332-1GEA 0 GBP
COBBAN-1332-1G COBBAN-1332-25G COBBAN-1332-5G
5-Fluoro-o-anisidine hydrochloride
5-Fluoro-o-anisidine hydrochloride
Formula: C₇H₉ClFNO
MDL Number: MFCD09997942
CAS Number: 326-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR