2,4,6-Tribromo-1-(trifluoromethylthio)benzene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-1103-1G OT-1103-250M
COBBOT-1103-1GEA 0 GBP
COBBOT-1103-1G COBBOT-1103-250M
2,4,6-Tribromo-1-(trifluoromethylthio)benzene
2,4,6-Tribromo-1-(trifluoromethylthio)benzene
Formula: C₇H₂Br₃F₃S
MDL Number: MFCD13195742
CAS Number: 1215205-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR