3-Bromo-2-chloroanisole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Bromo-2-chloro-3-methoxybenzene

OT-1170-5G OT-1170-1G
COBBOT-1170-5GEA 0 GBP
COBBOT-1170-5G COBBOT-1170-1G
3-Bromo-2-chloroanisole
3-Bromo-2-chloroanisole
Formula: C₇H₆BrClO
MW: 221.4789 g/mol
MDL Number: MFCD16036127
CAS Number: 174913-11-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR