6-Methylpicolinic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-Methyl-2-pyridinecarboxylic acid, 6-Methylpyridine-2-carboxylic acid

011322-5G 011322-25G
FLUO011322-5GEA 0 GBP
FLUO011322-5G FLUO011322-25G
6-Methylpicolinic acid
6-Methylpicolinic acid
Formula: C₇H₇NO₂
MW: 137.14 g/mol
MDL Number: MFCD00023481
CAS Number: 934-60-1
EINECS: 213-287-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR