2-Bromo-6-fluoro-3-methylphenol

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-1226-1G OT-1226-5G
COBBOT-1226-1GEA 0 GBP
COBBOT-1226-1G COBBOT-1226-5G
2-Bromo-6-fluoro-3-methylphenol
2-Bromo-6-fluoro-3-methylphenol
Formula: C₇H₆BrFO
MDL Number: MFCD16295100
CAS Number: 1226808-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR