2-Methoxy-6-(trifluoromethyl)pyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PY-7906-1G
PY-7906-5G
COBBPY-7906-1GEA
0
GBP
COBBPY-7906-1G
COBBPY-7906-5G
2-Methoxy-6-(trifluoromethyl)pyridine
2-Methoxy-6-(trifluoromethyl)pyridine
Formula:
C₇H₆F₃NO MW: 177.1200 g/mol |
MDL Number:
MFCD12026108 CAS Number: 34486-18-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...