4-Fluoro-5-methoxy-2-nitroaniline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-3276-5G HC-3276-1G
COBBHC-3276-5GEA 0 GBP
COBBHC-3276-5G COBBHC-3276-1G
4-Fluoro-5-methoxy-2-nitroaniline
4-Fluoro-5-methoxy-2-nitroaniline
Formula: C₇H₇FN₂O₃
MDL Number: MFCD00247494
CAS Number: 125163-12-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR