1-(2-(Trifluoromethyl)phenyl)thiourea

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: N-[2-(Trifluoromethyl)phenyl]thiourea

009389-5G 009389-25G
FLUO009389-5GEA 0 GBP
FLUO009389-5G FLUO009389-25G
1-(2-(Trifluoromethyl)phenyl)thiourea
1-(2-(Trifluoromethyl)phenyl)thiourea
Formula: C₈H₇F₃N₂S
MW: 220.22 g/mol
MDL Number: MFCD00041161
CAS Number: 1736-71-6
EINECS: 217-084-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR