3-(4-Fluorophenoxy)propionic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CFF625.1
001654-1G
001654-5G
CHMPCFF625.1EA
0
GBP
CHMPCFF625.1
FLUO001654-1G
FLUO001654-5G
3-(4-Fluorophenoxy)propionic acid
3-(4-Fluorophenoxy)propionic acid
Formula:
C₉H₉FO₃ |
MDL Number:
MFCD00052838 CAS Number: 1579-78-8 EINECS: 216-424-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...