3-(3-Fluorophenoxy)propionic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(3-Fluorophenoxy)propionic Acid

AS22923G1 AS22923G5
ACTIAS22923G1EA 0 GBP
ACTIAS22923G1 ACTIAS22923G5
3-(3-Fluorophenoxy)propionic acid
3-(3-Fluorophenoxy)propionic acid
Formula: C₉H₉FO₃
MW: 184.1644 g/mol
MDL Number: MFCD01931913
CAS Number: 133077-42-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR