3-(2-Fluorophenoxy)propionic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(2-Fluorophenoxy)propionic Acid

AS22797G1 AS22797G5
ACTIAS22797G1EA 0 GBP
ACTIAS22797G1 ACTIAS22797G5
3-(2-Fluorophenoxy)propionic acid
3-(2-Fluorophenoxy)propionic acid
Formula: C₉H₉FO₃
MW: 184.1600 g/mol
MDL Number: MFCD01569309
CAS Number: 2967-72-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR