3-(3-Methylphenoxy)propionic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22932G1 AS22932G5
ACTIAS22932G1EA 0 GBP
ACTIAS22932G1 ACTIAS22932G5
3-(3-Methylphenoxy)propionic acid
3-(3-Methylphenoxy)propionic acid
Formula: C₁₀H₁₂O₃
MDL Number: MFCD00642411
CAS Number: 25173-36-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR