3-(2-Methylphenoxy)propionic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS22933G1
AS22933G5
ACTIAS22933G1EA
94
GBP
ACTIAS22933G1
ACTIAS22933G5
3-(2-Methylphenoxy)propionic acid
3-(2-Methylphenoxy)propionic acid
Formula:
C₁₀H₁₂O₃ |
MDL Number:
MFCD00642410 CAS Number: 25173-35-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...