3-(4-Methoxyphenoxy)propionic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22924G1 AS22924G5
ACTIAS22924G1EA 0 GBP
ACTIAS22924G1 ACTIAS22924G5
3-(4-Methoxyphenoxy)propionic acid
3-(4-Methoxyphenoxy)propionic acid
Formula: C₁₀H₁₂O₄
MDL Number: MFCD00020515
CAS Number: 20811-60-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR