3-(2-Methoxyphenoxy)propionic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22926G1 AS22926G5
ACTIAS22926G1EA 129 GBP
ACTIAS22926G1 ACTIAS22926G5
3-(2-Methoxyphenoxy)propionic acid
3-(2-Methoxyphenoxy)propionic acid
Formula: C₁₀H₁₂O₄
MW: 196.1999 g/mol
MDL Number: MFCD01931914
CAS Number: 20370-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR