3-(4-Nitrophenoxy)propionic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(4-Nitrophenoxy)propanoic acid

AS22911G25 AS22911G5
ACTIAS22911G25EA 542 GBP
ACTIAS22911G25 ACTIAS22911G5
3-(4-Nitrophenoxy)propionic acid
3-(4-Nitrophenoxy)propionic acid
Formula: C₉H₉NO₅
MW: 211.1700 g/mol
MDL Number: MFCD00043748
CAS Number: 10572-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR