3-(3-Nitrophenoxy)propionic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22912G1 AS22912G5
ACTIAS22912G1EA 0 GBP
ACTIAS22912G1 ACTIAS22912G5
3-(3-Nitrophenoxy)propionic acid
3-(3-Nitrophenoxy)propionic acid
Formula: C₉H₉NO₅
MDL Number: MFCD09049926
CAS Number: 91004-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR