3-(2-Nitrophenoxy)propionic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22913G1 AS22913G5
ACTIAS22913G1EA 167 GBP
ACTIAS22913G1 ACTIAS22913G5
3-(2-Nitrophenoxy)propionic acid
3-(2-Nitrophenoxy)propionic acid
Formula: C₉H₉NO₅
MDL Number: MFCD09045698
CAS Number: 6336-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR