4-Aminopyridine-2-carbonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Aminopicolinonitrile

AS12345G1 AS12345G5
ACTIAS12345G1EA 131 GBP
ACTIAS12345G1 ACTIAS12345G5
4-Aminopyridine-2-carbonitrile
4-Aminopyridine-2-carbonitrile
Formula: C₆H₅N₃
MW: 119.12 g/mol
MDL Number: MFCD07374351
CAS Number: 98139-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR