3-(3-Chlorophenoxy)propionic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22927G1 AS22927G5
ACTIAS22927G1EA 127 GBP
ACTIAS22927G1 ACTIAS22927G5
3-(3-Chlorophenoxy)propionic acid
3-(3-Chlorophenoxy)propionic acid
Formula: C₉H₉ClO₃
MDL Number: MFCD00151673
CAS Number: 7170-50-5
EINECS: 230-522-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR