4-Bromo-N-isopropylaniline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AN-1413-5G
AN-1413-1G
COBBAN-1413-5GEA
0
GBP
COBBAN-1413-5G
COBBAN-1413-1G
4-Bromo-N-isopropylaniline
4-Bromo-N-isopropylaniline
Formula:
C₉H₁₂BrN MW: 214.1023 g/mol |
MDL Number:
MFCD08691756 CAS Number: 121086-19-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...