(1-Propynyl)tributyltin

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS50708G1 AS50708G25 AS50708G5
ACTIAS50708G1EA 0 GBP
ACTIAS50708G1 ACTIAS50708G25 ACTIAS50708G5
(1-Propynyl)tributyltin
(1-Propynyl)tributyltin
Formula: C₁₅H₃₀Sn
MDL Number: MFCD01863650
CAS Number: 64099-82-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR