4-(Difluoromethoxy)iodobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
004837-1G
004837-5G
004837-25G
FLUO004837-1GEA
0
GBP
FLUO004837-1G
FLUO004837-5G
FLUO004837-25G
4-(Difluoromethoxy)iodobenzene
4-(Difluoromethoxy)iodobenzene
Formula:
C₇H₅F₂IO |
MDL Number:
MFCD00085008 CAS Number: 128140-82-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...