3-(Dimethylamino)benzaldehyde

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(N,N-Dimethylamino)benzaldehyde

AS36160G1 AS36160G5
ACTIAS36160G1EA 0 GBP
ACTIAS36160G1 ACTIAS36160G5
3-(Dimethylamino)benzaldehyde
3-(Dimethylamino)benzaldehyde
Formula: C₉H₁₁NO
MW: 149.1897 g/mol
MDL Number: MFCD05148649
CAS Number: 619-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR