4-Fluoro-2-(trifluoromethyl)cinnamic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007932-5G 007932-1G
FLUO007932-5GEA 0 GBP
FLUO007932-5G FLUO007932-1G
4-Fluoro-2-(trifluoromethyl)cinnamic acid
4-Fluoro-2-(trifluoromethyl)cinnamic acid
Formula: C₁₀H₆F₄O₂
MDL Number: MFCD00236297
CAS Number: 243977-21-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR