5-Fluoro-2-(trifluoromethyl)cinnamic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
007934-1G
007934-5G
FLUO007934-1GEA
0
GBP
FLUO007934-1G
FLUO007934-5G
5-Fluoro-2-(trifluoromethyl)cinnamic acid
5-Fluoro-2-(trifluoromethyl)cinnamic acid
Formula:
C₁₀H₆F₄O₂ |
MDL Number:
MFCD00236299 CAS Number: 231291-18-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...