2-Fluoro-3-(trifluoromethyl)cinnamic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007926-1G 007926-5G
FLUO007926-1GEA 0 GBP
FLUO007926-1G FLUO007926-5G
2-Fluoro-3-(trifluoromethyl)cinnamic acid
2-Fluoro-3-(trifluoromethyl)cinnamic acid
Formula: C₁₀H₆F₄O₂
MDL Number: MFCD00236291
CAS Number: 237069-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR