1,4-Benzodioxane-2-carboxylic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,3-Dihydrobenzo[b][1,4]dioxine-2-carboxylic acid

AS20760G100 AS20760G25
ACTIAS20760G100EA 131 GBP
ACTIAS20760G100 ACTIAS20760G25
1,4-Benzodioxane-2-carboxylic acid
1,4-Benzodioxane-2-carboxylic acid
Formula: C₉H₈O₄
MW: 180.16 g/mol
MDL Number: MFCD00084888
CAS Number: 3663-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR