6-Methylisatin

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS24770G1 AS24770G5
ACTIAS24770G1EA 0 GBP
ACTIAS24770G1 ACTIAS24770G5
6-Methylisatin
6-Methylisatin
Formula: C₉H₇NO₂
MDL Number: MFCD01718832
CAS Number: 1128-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR